Learn More
Syngene™ ULPA Filter
GREENER_CHOICE
Product Code. 16514351 Shop All Syngene Products
Change view
Click to view available options
Filter Type:
ULPA
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Filter Type unitSize
16514351 ULPA Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16514351 Supplier Syngene™ Supplier No. SYN49FULPA
UPLA filter for use with PCR Workstation.
  • Spare ULPA filter
Specifications
Efficiency 99.99% Efficient at 0.125mu;
Filter Type ULPA