Learn More
Qsonica Ultrasonic Homogenizer Converters
Product Code. 15142889 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Product Type:
Converter Cable
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15142889 Converter Cable Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Product Code. 15142889
  • ONLINE EXCLUSIVE PRICE
    173.00€
    133.55€ / Pack of 1
    Save 39.45€
    22% Off
Estimated Shipment: 17-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Qsonica Ultrasonic Homogenizer Converter works with either the Microson XL2000 or the Sonicator 3000 Ultrasonic Homogenizers to transform electrical signals into mechanical vibrations.
  • For use with Microson™ Model XL2000 or Sonicator™ 3000 Ultrasonic Homgenizers
  • Transforms electrical signal into mechanical vibrations
Specifications
Product Type Converter Cable
Length (English) 72 in.
Length (Metric) 182.88 cm
For Use With (Application) Ideal for transform electrical signal into mechanical vibration
For Use With (Equipment) Q500 and Q700 Sonicator™