missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UMPS Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32172PCP
This item is not returnable.
View return policy
Description
UMPS Polyclonal antibody specifically detects UMPS in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| UMPS | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| EC 4.1.1.23, OMPdecase, OPRT, OPRTase, orotate phosphoribosyltransferase, orotidine 5'-phosphate decarboxylase, UMP synthase, uridine 5'-monophosphate synthase, uridine monophosphate synthetase | |
| The immunogen this antibody was made to, maps to a region between residue 430 to 480 of human Uridine Monophosphate Synthetase using the numbering given in entry NP_000364.1 | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing, metabolism | |
| 7372 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| PerCP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction