missing translation for 'onlineSavingsMsg'
Learn More

UNC0646, TRC

Product Code. 30455568
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30455568 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30455568 Supplier Toronto Research Chemicals Supplier No. TRCU7125105MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(1-Cyclohexyl-4-piperidinyl)-2-[hexahydro-4-(1-methylethyl)-1H-1,4-diazepin-1-yl]-6-methoxy-7-[3-(1-piperidinyl)propoxy]-4-quinazolinamine
CAS 1320288-17-2
Molecular Formula C14 H15 N7 . C4 H7 N O3
Quantity 5 mg
Synonym N-(1-Cyclohexylpiperidin-4-yl)-2-(4-isopropyl-1,4-diazepan-1-yl)-6-methoxy-7-(3-(piperidin-1-yl)propoxy)quinazolin-4-amine, UNC0646
InChI Formula InChI=1S/C14H15N7.C4H7NO3/c15-13(16)9-1-5-11(6-2-9)19-21-20-12-7-3-10(4-8-12)14(17)18;1-3(6)5-2-4(7)8/h1-8H,(H3,15,16)(H3,17,18)(H,19,20);2H2,1H3,(H,5,6)(H,7,8)
SMILES CC(=O)NCC(=O)O.NC(=N)c1ccc(N\N=N\c2ccc(cc2)C(=N)N)cc1
IUPAC Name 2-acetamidoacetic acid;4-[(2E)-2-(4-carbamimidoylphenyl)iminohydrazinyl]benzenecarboximidamide
Molecular Weight (g/mol) 398.419
Formula Weight 398.181
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.