missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UNC5A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22068-1-AP-150UL
This item is not returnable.
View return policy
Description
The UNC5H family of proteins act as transmembrane receptors for netrin-1 and play a crucial role in axon guidance and migration of neural cells. Additionally, when cleaved by a caspase to produce an intracellular fragment containing a death domain,UNC5H receptors induce apoptosis. This activity is blocked by the binding of netrin-1. In the absence of netrin-1, UNC5H receptors act as tumor suppressors by inhibiting anchorage-independent growth and invasion, but mutation of these receptors provides a potential mechanism for tumorigenicity. The expression of UNC5H receptors is downregulated in multiple carcinomas, including colorectal, breast, ovary, uterus, stomach, lung and kidney cancers. UNC5H1, also known as UNC5HA (unc-5 homolog A), is a member of the UNC5H family of proteins and is localized to the cell membrane. Three isoforms of UNC5H1 exist due to alternative splicing events.Specifications
| UNC5A | |
| Polyclonal | |
| Unconjugated | |
| UNC5A | |
| KIAA1976, Netrin receptor UNC5A, Protein unc 5 homolog 1, Protein unc 5 homolog A, unc 5 homolog A (C. elegans), UNC5A, UNC5H1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 107448, 60629, 90249 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O08721, Q6ZN44, Q8K1S4 | |
| Unc5a | |
| UNC5A Fusion Protein Ag17155 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction