Learn More
Invitrogen™ UNC5C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114231
Description
Immunogen sequence: EVSIEISRQQ VEELFGPEDY WCQCVAWSSA GTTKSRKAYV RIAYLRKTFE For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
This gene product belongs to the UNC-5 family of netrin receptors. Netrins are secreted proteins that direct axon extension and cell migration during neural development. They are bifunctional proteins that act as attractants for some cell types and as repellents for others, and these opposite actions are thought to be mediated by two classes of receptors. The UNC-5 family of receptors mediate the repellent response to netrin; they are transmembrane proteins containing 2 immunoglobulin -like domains and 2 type I thrombospondin motifs in the extracellular region.
Specifications
| UNC5C | |
| Polyclonal | |
| Unconjugated | |
| Unc5c | |
| B130051O18Rik; Netrin receptor UNC5C; protein unc-5 homolog 3; Protein unc-5 homolog C; Rcm; Rostral cerebellar malformation protein; unc5 (C.elegans homolog) c; unc5 homolog 3; UNC-5 homolog 3; unc-5 homolog C; unc-5 homolog C (C. elegans); unc-5 netrin receptor C; UNC5C; UNC5H3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 8633 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O95185 | |
| Unc5c | |
| Synthetic peptide directed towards the middle region of Human UNC5C. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.