missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UNC5C Polyclonal Antibody

Brand: Thermo Scientific PA5114788
This item is not returnable.
View return policy
Description
Specifications
| UNC5C | |
| Polyclonal | |
| Unconjugated | |
| Unc5c | |
| B130051O18Rik; Netrin receptor UNC5C; protein unc-5 homolog 3; Protein unc-5 homolog C; Rcm; Rostral cerebellar malformation protein; unc5 (C.elegans homolog) c; unc5 homolog 3; UNC-5 homolog 3; unc-5 homolog C; unc-5 homolog C (C. elegans); unc-5 netrin receptor C; UNC5C; UNC5H3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 22253, 362049, 8633 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O08747, O95185, Q761X5 | |
| Unc5c | |
| A synthesized peptide derived from human UNC5C(Accession O95185), corresponding to amino acid residues K84-L134. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction