missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UNC84A Recombinant Rabbit Monoclonal Antibody (24GB2575)

Rabbit Recombinant Monoclonal Antibody

198.00€ - 564.00€

Specifications

Antigen UNC84A
Clone 24GB2575
Concentration 0.33 mg/mL
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30276658
View Documents
Invitrogen™
MA557748
100 μL
564.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30274901
View Documents
Invitrogen™
MA557749
20 μL
198.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

UNC84A Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

This gene is a member of the unc-84 homolog family and encodes a nuclear envelope protein with an Unc84 (SUN) domain. The protein is involved in nuclear anchorage and migration. Several alternatively spliced transcript variants of this gene have been described; however, the full-length nature of some of these variants has not been determined.
Specifications

Specifications

UNC84A
0.33 mg/mL
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O94901
23353
A synthesized peptide derived from human SUN1 (aa 110-167).
Antibody
24GB2575
-20°C
Recombinant Monoclonal
Liquid
IgG
Human
SUN1
4632417G13Rik; 5730434D03Rik; KIAA0810; mKIAA0810; Protein unc-84 homolog A; Sad1 and UNC84 domain containing 1; Sad1 unc-84 domain protein 1; sad1/unc-84 protein-like 1; SUN domain-containing protein 1; Sun1; unc-84 homolog A; Unc84a
SUN1
Primary
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.