Learn More
Abeba™ UNI6 36780 Shoes
Product Code. 15650638
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15650638 35 1 pair
15660638 36 1 pair
15670638 37 1 pair
15680638 38 1 pair
15690638 39 1 pair
15600648 40 1 pair
15610648 41 1 pair
15620648 42 1 pair
15630648 43 1 pair
15640648 44 1 pair
15650648 45 1 pair
15660648 46 1 pair
15670648 47 1 pair
15680648 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 15650638 Supplier Abeba™ Supplier No. 36780/35
Only null left
Add to basket
Add to basket
ESD dissipative shoe includes special lining with silver ions that absorbs resperation which allows the foot to breathe.
  • CE EN ISO 20347:2012 O2 FO SRC
  • Microfiber
  • White
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable Soft Comfort insole M (article no. 350112)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
Specifications
Certifications/Compliance CE EN ISO 20347:2012 O2 FO SRC
Color White
Material Microfiber
Size 35
Sole Features Slip-resistant
Sole Material Polyurethane
Style 36780