Learn More
Cytiva (Formerly GE Healthcare Life Sciences) UNICORN start
Product Code. 17059203 Shop All Cytiva Products
Change view
Click to view available options
Includes:
4 modules
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
17059203 4 modules Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17059203 Supplier Cytiva (Formerly GE Healthcare Life Sciences) Supplier No. 29480207
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Control software designed for ÄKTA start
  • Simple and flexible method creation using predefined templates and phases (steps)
  • Enables real-time monitoring of the run and intuitive way to control the run
  • Evaluate, compare, and share results
  • Create and print PDF reports
  • Robust database storage with easy access to data, data security, and data integrity
  • Maximizes performance
  • Includes:
    • Four different modules:
      • System control
      • Method editor
      • Evaluation
      • Administration
    TRUSTED_SUSTAINABILITY
    Specifications
    Type Control Software
    Description DVD without license
    For Use With (Equipment) Windows™ 7 Professional SP1 (US English 32-Bit and 64-Bit), Windows™ 10 Professional (US English 32-Bit and 64-Bit)
    Includes 4 modules
    Name des Produkts
    Wählen Sie ein Thema
    Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.