Description
- Hand-operated vacuum pump
- No electricity, no additional equipment required
- Contamination-proof sample guidance, contact only with original bottle and tube
- Universal bottle adapter for various bottle necks (Ø 24-50 mm) and sample bottles (100-1000 mL)
- Ready for immediate use
- Including sample bottles and label seals for secure documentation
Specifications
Specifications
| Product Type | Liquid Sampler |
| Material | Polypropylene |
| Autoclavable | Not Autoclavable |
| Includes | Suction lance, 5 sample bottles each PP 250 mL and 500 mL, Vacuum pump, Universal adapter for sample bottles, 20 seal-it bottle seals |
| Capacity (English) | 8.45 oz. |
| Capacity (Metric) | 250 mL |
| For Use With (Application) | For Liquid Samples From Precisely Pre-Determined Depth, with Hand-Operated Vacuum Pump, Target sample |
| Length (Metric) Blade | 100 cm |