Learn More
Unity Lab Services™ Unity Basic Plan Autoclave Group 1
Maximize technology investment by predicting and preventing problems before they occur.
Brand: Unity Lab Services™ ULS_BASIC_ATC-1
Additional Details : Weight : 0.00010kg
Description
Basic Support Plan Features:
- Priority status for technical support inquiries
- One (1) Scheduled Preventative Maintenance visit*
- 25% discount on parts required for service calls
- 10% discount on labor and travel required for service calls
- Software updates and notifications
*Parts required for PM visit are not included but 25% discount applies
Specifications
Autoclave Group 1 | |
Basic Plan |