Learn More
IKA™ Universal Batch Mill
Product Code. 10111361 Shop All IKA Products
Change view
Click to view available options
Capacity (Metric):
250 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10111361 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Product Code. 10111361 Supplier IKA™ Supplier No. 0001603600
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Grinds hard and brittle substances up to 250 ml with a fixed speed of 20000 rpm. IKA™ Universal Batch Mill has two grinding chambers that can be alternately operated using one drive. M 21 blade included with delivery.
Batch mill suitable for dry grinding of hard and brittle substances up to 250mL with a fixed speed of 20,000rpm.
  • Double-walled grinding chamber can be cooled with water through two hose adapters
  • Removable grinding chamber is easy to clean
  • Two grinding chambers can be alternately operated using one drive
  • M 21 blade included
TRUSTED_SUSTAINABILITY
Specifications
Product Type Mill
Capacity (Metric) 250 mL
Certifications/Compliance IP 21, DIN EN 60529
Depth (Metric) 170 mm
Electrical Requirements 230/115 V, 50/60 Hz, 550 W
For Use With (Equipment) For Dry Grinding of Hard and Brittle Substances, Two Grinding Chambers
Hardness Max: 5 Mohs
Height (Metric) 350 mm
Material Stainless Steel
Speed 20,000 rpm
Voltage 230/115 V
Watts 550 W
Width (Metric) 170 mm
Frequency 50/60 Hz