Learn More
Vitlab™ Universal Model Natural Rubber Pipet Filler
Product Code. 10499382
Change view
Click to view available options
Type:
Pipette Filler
For Use With (Equipment):
For pipettes up to 100mL
Material:
Natural Rubber
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Equipment) Material unitSize
10499382 Pipette Filler For pipettes up to 100mL Natural Rubber Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10499382 Supplier Vitlab™ Supplier No. 104199
In Stock
Add to basket
Add to basket
Manufactured from natural rubber. Vitlab™ Universal Model Natural Rubber Pipet Filler is used for pipetting with volumetric or measuring pipettes.
  • For pipetting with volumetric or measuring pipettes
  • With 3 valves:
    • Valve A: Air release
    • Valve B: Liquid filling
    • Valve E: Liquid dispensing
Specifications
Material Natural Rubber
Quantity 1 Each
Type Pipette Filler
For Use With (Equipment) For pipettes up to 100mL