Learn More
Kartell™ Plastilab Untapered T-Shaped Tubing Connectors
Click to view available options
Port Size:
1.6mm bore
3.7mm
4.4mm bore
6.3mm bore
Packaging Quantity:
10/Pk.
100/Pk.
Unit Size:
Pack of 10
Pack of 100
Description
Designed with serrations to hold hoses tightly and prevent slipping.
- Made from polypropylene
- Autoclavable
- Chemical- and temperature-resistant
Compliance
Meet Food and Agriculture regulations
Specifications
Specifications
| Certifications/Compliance | Meet with the Food and Drug regulations. 90/128/CEE 92/39/CEE e FDA 21 CFR 177.1520, CE and IVD |
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| For Use With (Application) | Suitable for foodstuff |
| Packaging Quantity | 100/Pk. |
| Port Size | 3.7mm |
| Shape | T-shape |
| Product Type | Untapered T Tubing Connector |