missing translation for 'onlineSavingsMsg'
Learn More

UPF3B Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA541903

Additional Details : Weight : 0.00010kg

Product Code. 15525122

  • 472.00€ / 100µL
Estimated Shipment: 27-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Peptide sequence: MKEEKEHRPK EKRVTLLTPA GATGSGGGTS GDSSKGEDKQ DRNKEKKEAL Sequence homology: Cow: 100%; Dog: 100%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 93%; Rat: 100%

UPF3B Polyclonal antibody specifically detects UPF3B in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
Specifications

Specifications

UPF3B
Polyclonal
Unconjugated
UPF3B
5730594O13Rik; AI317193; AW541158; HUPF3B; hUpf3p-X; mental retardation, X-linked 62; MRX62; MRXS14; Nonsense mRNA reducing factor 3B; regulator of nonsense transcripts 3B; RENT3B; RGD1560264; Unknown (protein for MGC:134301); UPF3 regulator of nonsense transcripts homolog B; UPF3 regulator of nonsense transcripts homolog B (yeast); UPF3 regulator of nonsense transcripts-like protein B; UPF3B; UPF3B pseudogene 1; UPF3B pseudogene 2; UPF3B pseudogene 3; UPF3B regulator of nonsense mediated mRNA decay; UPF3BP1; UPF3BP2; UPF3BP3; Upf3p-X; UPF3X; up-frameshift suppressor 3 homolog B; Up-frameshift suppressor 3 homolog on chromosome X
Rabbit
Protein A
RUO
65109
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9BZI7
UPF3B
Synthetic peptide directed towards the N-terminal of human UPF3B
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions