missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UQCRC2 Recombinant Rabbit Monoclonal Antibody (24GB2220)

Rabbit Recombinant Monoclonal Antibody
198.00€ - 564.00€
Specifications
| Antigen | UQCRC2 |
|---|---|
| Clone | 24GB2220 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30276043
![]() |
Invitrogen™
MA557828 |
100 μL |
564.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275572
|
Invitrogen™
MA557829 |
20 μL |
198.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UQCRC2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. The core protein 2 is required for the assembly of the complex.Specifications
| UQCRC2 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Rat | |
| UQCRC2 | |
| 1500004O06Rik; complex III subunit 2; Core protein II; cytochrome bc-1 complex core protein II; cytochrome b-c1 complex subunit 2, mitochondrial; MC3DN5; QCR2; U MC3DN5; ubiquinol cytochrome c reductase core protein 2; ubiquinol-cytochrome C reductase complex core protein 2, mitochondrial precursor (Complex III subunit II); ubiquinol-cytochrome c reductase core protein II; ubiquinol-cytochrome-c reductase complex core protein 2; UQCR2; Uqcrc2 | |
| UQCRC2 | |
| Primary | |
| Affinity chromatography |
| 24GB2220 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P22695, P32551 | |
| 293448, 7385 | |
| A synthesized peptide derived from human UQCRC2 (aa 300-375). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title