Learn More
URB2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24881-1-AP-150UL
This item is not returnable.
View return policy
Description
URB2 gene ontology annotations related to this gene include ribosome biogenesis.Specifications
| URB2 | |
| Polyclonal | |
| Unconjugated | |
| URB2 | |
| KIAA0133, NET10, URB2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9816 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q14146 | |
| URB2 | |
| URB2 Fusion Protein Ag21667 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |