missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Urea Broth (Christensen and Maslen)

Product Code. 11994042
Change view
Click to view available options
Fill Weight:
5.5 to 6.5 g
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Weight Unit Size
11994042 5.5 to 6.5 g Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11994042 Supplier Thermo Scientific™ Supplier No. TV5007N
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detect urease activity in bacteria by observing a color change due to the hydrolysis of urea, which helps differentiate urease- producing Enterobacteriaceae from members of the Salmonella and Shigella groups with this medium.

Urea Broth (Christensen and Maslen) is used to detect urease activity in bacteria by observing a color change due to the hydrolysis of urea, which helps differentiate urease- producing Enterobacteriaceae from members of the Salmonella and Shigella groups with this medium.

TRUSTED_SUSTAINABILITY

Specifications

Color Pastel Yellow, Transparent
Content And Storage 2°C to 25°C
Format Poured Tube
Manufacturing Origin Germany
Product Type Microbiology Media
Fill Weight 5.5 to 6.5 g
Packaging Type 50 Tubes/Box
Quantity 50/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.