missing translation for 'onlineSavingsMsg'
Learn More

Ursocholic Acid, TRC

Product Code. 30008610
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008610 25 mg 25mg
30005189 1 mg 1mg
30010436 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008610 Supplier Toronto Research Chemicals Supplier No. TRCU84990025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2955-27-3
Molecular Formula C24 H40 O5
Molecular Weight (g/mol) 408.57
Synonym (3alpha,5beta,7beta,12alpha)-3,7,12-Trihydroxycholan-24-oic acid, 3alpha,7beta,12alpha-Trihydroxy-5beta-cholan-24-oic acid, 3alpha,7beta,12alpha-Trihydroxy-5beta-cholanic acid, 3alpha,7beta,12alpha-Trihydroxy-5beta-cholanoic acid, 3alpha,7beta,12alpha-Trihydroxycholanic acid, 7-Epicholic acid, 7beta-Hydroxyisocholic acid, Ursocholic acid
IUPAC Name (4R)-4-[(3R,5S,7S,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
SMILES C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C24H40O5/c1-13(4-7-21(28)29)16-5-6-17-22-18(12-20(27)24(16,17)3)23(2)9-8-15(25)10-14(23)11-19(22)26/h13-20,22,25-27H,4-12H2,1-3H3,(H,28,29)/t13-,14+,15-,16-,17+,18+,19+,20+,22+,23+,24-/m1/s1
Formula Weight 408.2876
Percent Purity >95%
Chemical Name or Material Ursocholic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.