Learn More
USP36 Monoclonal Antibody (OTI9F5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800098
Description
May be required for maintaining multiple types of adult stem cells. May function as a transcriptional repressor by continually deubiquiting histone H2B at the promoters of genes critical for cellular differentiation, thereby preventing histone H3 ′Lys-4′ trimethylation (H3K4). Nagase T., DNA Res. 7:143-150(2000). The MGC Project Team, Genome Res. 14:2121-2127(2004). Ota T., Nat. Genet. 36:40-45(2004).Specifications
| USP36 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9P275 | |
| USP36 | |
| Human recombit protein fragment corresponding to amino acids 589-972 of human USP36 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI9F5 | |
| Unconjugated | |
| USP36 | |
| DUB1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 57602 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.