missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRMT5 Antibody (OTI1B5), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44733
This item is not returnable.
View return policy
Description
TRMT5 Monoclonal antibody specifically detects TRMT5 in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| TRMT5 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized powder ( 1X PBS, pH 7.3,) | |
| Mouse | |
| >95% | |
| RUO | |
| 57570 | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry, Western Blot | |
| OTI1B5 | |
| Western Blot 1:2000, Immunohistochemistry 1:1000 | |
| KIAA1393EC 2.1.1.31, M1G-methyltransferase, TRM5 tRNA methyltransferase 5 homolog (S. cerevisiae), TRM5MGC111453, tRNA (guanine-N(1)-)-methyltransferase, tRNA [GM37] methyltransferase, tRNA methyltransferase 5, tRNA-N1G37 methyltransferase | |
| Human recombinant protein fragment corresponding to amino acids 1-303 of human TRMT5(NP_065861) produced in E.coli. | |
| 100 μg | |
| Primary | |
| For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process. | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction