Learn More
USP9X Monoclonal Antibody (OTI2B4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800061S
Description
USP9X Monoclonal antibody specifically detects USP9X in Human samples. It is validated for Western BlotSpecifications
| USP9X | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q969T4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10477 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2B4 | |
| Unconjugated | |
| UBE2E3 | |
| 5730589N07Rik; AA407302; AA407699; AL022658; AL022749; Deubiquitinating enzyme FAF-X; DFFRX; Drosophila fat facets related, X-linked; FAF; Fafl; FAF-X; FAM; fat facets homolog; Fat facets in mammals; fat facets protein related, X-linked; fat facets protein-related, X-linked; fats facets protein related, X; hFAM; MRX99; MRXS99F; Probable ubiquitin carboxyl-terminal hydrolase FAF-X; putative ubiquitin carboxyl-terminal hydrolase FAF-X; putative ubiquitin carboxyl-terminal hydrolase FAF-x-like protein; RGD1560056; Ubiquitin carboxyl-terminal hydrolase FAM; ubiquitin specific peptidase 9 X-linked; ubiquitin specific peptidase 9, X chromosome; ubiquitin specific peptidase 9, X-linked; ubiquitin specific protease 9 SSSRF- isoform; ubiquitin specific protease 9, X chromosome; ubiquitin specific protease 9, X chromosome (fat facets-like Drosophila); ubiquitin specific protease 9, X-linked; ubiquitin thioesterase FAF-X; ubiquitin thiolesterase FAF-X; ubiquitin-specific processing protease FAF-X | |
| Full length human recombinant protein of human UBE2E3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.