Learn More
USP9X Monoclonal Antibody (OTI2B4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800061
Description
USP9X is an ubiquitin-specific protease involved in the removal and recycling of ubiquitin from proteins degraded in proteasomes. As a deubiquitinating enzyme, USP9x has a critical function in tight junction biogenesis by controlling the levels of the exchange factor for Arf6 (EFA6), a protein shown to facilitate tight junction formation during a narrow temporal window preceding the establishment of cell polarity (1). USP9X is also located on the X-chromosome but escapes X-inactivation, and defects in USP9X may cause gonadal degeneration in Turner syndrome.Specifications
| USP9X | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q93008 | |
| USP9X | |
| Human recombit protein fragment corresponding to amino acids 2246-2570 of human USP9X produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2B4 | |
| Unconjugated | |
| USP9X | |
| DFFRX, FAF, FAM, MRX99, MRXS99F | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8239 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.