Learn More
ERLAB UV Decontamination
Product Code. 30484797 Shop All Erlab Products
Change view
Click to view available options
Voltage:
0/1 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
30484797 0/1 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30484797 Supplier ERLAB Supplier No. NORT000475
  • ONLINE EXCLUSIVE PRICE
    312.00€
    290.00€ / Pack of 1
    Save 22.00€
    7% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Installed in microbiological safety cabinets, provides an efficient method for sterilizing the workspace
  • UV Decontamination uses germicidal UV-C light to destroy microorganisms on surfaces within the cabinet
  • Automatically or manually operated for timed decontamination cycles between work sessions
  • Promotes aseptic conditions for sensitive microbiological or cell culture applications
  • Reduces the need for harsh chemical disinfectants inside the workspace
  • Integrated safety sensors disable UV light when the sash is open, protecting users from exposure
  • Recommended for daily sterilization routines in biosafety environments
  • UV lamp is easy to replace and provides consistent performance over time
Specifications
Voltage 0/1 V