Learn More
Cytiva (Formerly GE Healthcare Life Sciences) UV Flow Cell i.d. 1 in.
Product Code. 16824953 Shop All Cytiva Products
Change view
Click to view available options
Length (Metric):
2.54 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
16824953 2.54 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16824953 Supplier Cytiva (Formerly GE Healthcare Life Sciences) Supplier No. 28959578
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Cytiva UV flow cell 1 in.
TRUSTED_SUSTAINABILITY
Specifications
Description Does not include TC optical fibers
Material Fluorinated Propylene Monomer (FPM/FKM), Polyetheretherketone (PEEK)
Length (Metric) 2.54 cm