Learn More
Cytiva UV Module
Product Code. 15279774 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
ÄKTA Start
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15279774 ÄKTA Start Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15279774 Supplier Cytiva Supplier No. 29024018
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use in protein purification applications
The AKTA start protein purification system is equipped with a UV monitor to continuously measure the absorbance of the liquid in the UV flow cell at a fixed wavelength (280nm). The UV monitor incorporates durable and reliable LED technology that is ready-to-use without any warm-up time. Moreover, the UV monitor does not generate any local heating of the flow cell, making it particularly suitable for heat-sensitive samples. The monitor is available with a 2mm optical path length flow cell and gives linear absorbance up to 1.5 AU.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) ÄKTA Start
Quantity 1 Each