Learn More
Fisherbrand™ UV Trays
Product Code. 11863353 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Dimensions (L x W):
10 x 150 mm
100 x 70 mm
100mm x 100mm
200mm x 100mm
200mm x 200mm
200mm x 250mm
70mm x 100mm
70mm x 150mm
70mm x 70mm
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
11863353 100mm x 100mm Each
11803423 70mm x 150mm Each
11883413 10 x 150 mm Each
11813473 200mm x 100mm Each
11833473 200mm x 250mm Each
11823473 200mm x 200mm Each
11837573 70mm x 100mm Each
11847573 70mm x 70mm Each
11873353 100 x 70 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 11863353
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
Accessory for Fisherbrand Horizontal Gel Electrophoresis Systems.
  • Accessory for Fisherbrand Horizontal Gel Electrophoresis Systems
Specifications
For Use With (Equipment) Sub-Gel Midi Horizontal Gel Electrophoresis System
Type UV Tray
Dimensions (L x W) 100mm x 100mm