Learn More
UVP Replacement UV Light Tubes
Product Code. 12883918
Change view
Click to view available options
Wavelength:
302 nm
365 nm
Wattage:
6 W
15 W
20 W
40 W
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wavelength Wattage unitSize
12883918 365 nm 15 W Each
12842989 302 nm 6 W Each
12813858 365 nm 40 W Each
12893848 365 nm 20 W Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12883918 Supplier UVP Supplier No. 34000902
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Replacement UV light tubes for 4, 6, 15, 20 and 40 watt ultraviolet lamps and transilluminators
Specifications
Wattage 15 W
Wavelength 365 nm
For Use With (Equipment) Model XX-15L UV bench lamp, ChromaDoc-It™ imaging system and Chromato-Vue™ C-70G, C-71, C-75 large UV viewing cabinets
Packaging Quantity 1 Ea.