Learn More
Greiner Bio-One Vacuette™ K2E K2 EDTA Tubes
Product Code. 15452520
Change view
Click to view available options
Capacity (Metric):
3 mL
Closure Color:
Black Ring
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Color unitSize
15452520 3 mL Black Ring Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15452520 Supplier Greiner Bio-One Supplier No. 454020
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Greiner Bio-One Vacuette™ K2E K2 EDTA Tubes is used for examination of whole blood in haematology.
  • For examination of whole blood in haematology
  • EDTA binds calcium ions and therefore blocks coagulation cascade
TRUSTED_SUSTAINABILITY
Specifications
Product Type K2 EDTA Tube
Capacity (Metric) 3 mL
Closure Color Black Ring
Tube Size 13 x 75 mm
compliance-icons
Product Identifier
  • X50 Tube  Premium K2E E.D.T.A. K2, 3 ml, 13x75,
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Ethylenediaminetetraacetic Acid Dipotassium Dihydrate