missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One VACUETTE™ Z No Additive Blood Collection Tubes

Product Code. p-9393003
Change view
Click to view available options
Capacity (Metric):
4 mL
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15432520 4 mL Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15432520 Supplier Greiner Bio-One Supplier No. 454001

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VACUETTE™ Z No Additive Collection Tubes are used in determinations for clinical chemistry and hormones, microbiological serology, immunology, TDM and crossmatching. These white top no additive blood tubes are typically used as discard tubes or as an alternative to serum tubes with a clot activator. They contain no chemical additives, are evacuated and sterile.

  • Tubes are supplied in packs of 50 pieces
  • For venous blood collection
  • Evacuated and are sterile
  • Made out of virtually unbreakable PET plastic
  • They provide improved safety and hygiene during specimen collection procedures with the advantage of being as clear as glass
  • Contain no additive in the tube. After one hour the blood in the tube is completely coagulated
  • CE marked
TRUSTED_SUSTAINABILITY

Specifications

Product Type Blood Collection Tube
Additive No Additive
Sterility Sterile
Closure Type Premium Safety Twist Cap
Capacity (Metric) 4 mL
Diameter (Metric) Inner 13 mm
For Use With (Equipment) VACUETTE™ Blood Collection Needles
Closure Color White
Height (Metric) 75 mm
Tube Size 13 x 75 mm
For Use With (Application) Blood collection
Length (Metric) 75 mm
Diameter (Metric) Outer 13 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.