Learn More
BD Vacutainer™ Hemogard™ Closure Plastic K3-Edta Tube
Product Code. 15349700
Change view
Click to view available options
Capacity (Metric):
2 mL
4 mL
Unit Size:
Pack of 100
Pack of 1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15349700 2 mL Pack of 100
12967666 2 mL Pack of 1000
12987666 4 mL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15349700 Supplier BD Supplier No. 367836
  • ONLINE EXCLUSIVE PRICE
    22.40€
    19.89€ / Pack of 100
    Save 2.51€
    11% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Use with whole blood or EDTA plasma sample. BD Vacutainer™ Hemogard™ Closure Plastic K3-EDTA Tube contains the additive K3 Potassium Salt of EDTA that acts as a coagulant.
  • Lavender Hemogard™ Closure
  • Used for whole blood or EDTA plasma sample
  • The additive K3 Potassium Salt of EDTA is a liquid in the Vacutainer Tube and acts as a coagulant
  • Available in sizes 2ml, 3ml and 4ml.
  • The difference between K2EDTA and K3EDTA is that the K3 is a liquid solution in the tubes, whereas the K2 is a liquid solution that is spray coated onto the tubes
TRUSTED_SUSTAINABILITY
Specifications
Product Type K2 EDTA Tube
Additive K3 EDTA Additive
Material Plastic
Closure Type Hemogard
Capacity (Metric) 2 mL
Label Type Block
Diameter (Metric) Outer 13 mm
Certifications/Compliance ICSH (International Council Society of Haematology)
compliance-icons
Product Identifier
  • X100 PLASTIC TUBE K3EDTA 2ML
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Tripotassium EDTA Dihydrate