missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ PET EDTA Tubes

Product Code. 12824811
Change view
Click to view available options
Material:
PET
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
12824811 PET Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12824811 Supplier BD Supplier No. 368856
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Product Type Vacutainer EDTA Tube
Additive K2 EDTA
Material PET
Closure Type Hemogard
Capacity (Metric) 3 mL
Closure Color Lavender
Tube Size 13 x 75 mm
For Use With (Application) Whole blood or EDTA plasma sample collection
Label Type Paper
compliance-icons
Product Identifier
  • X1000 BD Vacutainer EDTA tube
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: dipotassium EDTA dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.