Learn More
BD Vacutainer™ Venous Blood Collection Tubes: SST™ Serum Separation Tubes
Product Code. 15866161
Change view
Click to view available options
Capacity (Metric):
10 mL
Closure Color:
Red/Gray
Additive:
Polymer Gel/Silica Activator
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Color Additive Unit Size
15866161 10 mL Red/Gray Polymer Gel/Silica Activator Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15866161 Supplier BD Supplier No. 367985
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Ideal for use in serum determinations in chemistry
  • Contain spray-coated silica and a polymer gel for serum separation
  • Provide an efficient means of serum sample preparation and help to improve laboratory workflow
  • Tubes marked as Best Practice Formulary meet demanding requirements of complex diagnostic instruments and current clinical guidelines
  • Available with gold conventional tube stopper or red/gray Hemogard™ tube closure that prevents human contact with blood on the stopper or tube rim
Evacuated tubes simplify blood collection
  • Blood is drawn directly from the vein into the evacuated sterile collection tubes
  • Only one venipuncture is needed to draw multiple samples
  • Tubes can be changed while needle and plastic holder remain in place on patient's arm
  • Closures are color coded
Plastic Evacuated Tubes
  • Made of polyethylene terephthalate (PET)
  • Provide a safer work environment for all personnel by (1) reducing risk of tube breakage during handling; (2) eliminating breakage when tubes are centrifuged at higher speeds; and (3) eliminating breakage or chipping of tube rims when stopper is removed and reinserted
  • In addition to reducing potential injuries, fewer tube breakages result in less workflow interruption and fewer patient redraws, providing smoother day-to-day operation
Hemogard™ Stoppers
  • Safety shield designed for safer tube handling
  • Rubber stopper is recessed inside the plastic shield, so any drops left by a blood collection needle remain isolated from potential contact
  • Shield is designed to protect lab personnel from direct contact with blood on the stopper or around the outer rim of the tube, as well as from blood splattering upon opening the tube
TRUSTED_SUSTAINABILITY
Specifications
Additive Polymer Gel/Silica Activator
Material Plastic
Closure Type Traditional Cap
Capacity (Metric) 10 mL
Closure Color Red/Gray
Tube Size 16 x 125 mm
For Use With (Application) For use in serum determinations in chemistry