Learn More
Day-Impex Vacuum-insulated Stainless Steel Dewar Flasks
Product Code. 11783619
Product Code. 11783619 Supplier Day-Impex Supplier No. 320003
  • ONLINE EXCLUSIVE PRICE
    612.00€
    501.65€ / Pack of 1
    Save 110.35€
    18% Off
Estimated Shipment: 29-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Dilvac™ vacuum-insulated flasks are entirely stainless steel. Lightweight and compact in design they are suitable for ultra-low or very high temperature applications. Available in a range of capacities from 0.5 L to 6 L.
  • Vacuum-insulated
  • Lightweight stainless steel
  • Ideal for storage or transportation
  • Suitable for ultra-low and very high temperatures