Learn More
Welch Ilmvac™ Vacuum Pump Oil
Product Code. 10420853 Shop All Welch Ilmvac Products
Change view
Click to view available options
Volume (Metric):
5 L
20 L
Unit Size:
20 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10420853 20 L 20 L
10777544 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10420853 Supplier Welch Ilmvac™ Supplier No. 800124
  • ONLINE EXCLUSIVE PRICE
    352.00€
    320.30€ / 20 L
    Save 31.70€
    9% Off
View all promotions
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
One and two-stage rotary vane pump. Fisherbrand™ Vacuum Pump Oil designed for use with the range of Fisherbrand two-stage rotary vane pumps and with most other one and two-stage rotary vane pumps.
  • Generic mineral oil
  • Use in most one and two stage rotary vane pumps
  • 20L drum
  • Density: 0.888g/mL
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vacuum Pump Oil
Pressure 10 - 5 mbar
Viscosity 118 cSt viscosity at 40°C
Volume (Metric) 20 L