Learn More
Thermo Scientific™ High Pressure Stainless Steel Nuts and Ferrules
Product Code. 10446934 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Ferrule
Male Nut
Two-piece Fingerlok Nut with PEEK Ferrule
Valco 10-32 male hex nut
Quantity:
1 Each
10 Pack
Unit Size:
Each
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Quantity Unit Size
10446934 Male Nut 10 Pack Pack of 10
10639865 Two-piece Fingerlok Nut with PEEK Ferrule 1 Each Each
10423305 Valco 10-32 male hex nut 10 Pack Pack of 10
10170983 Ferrule 10 Pack Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10446934 Supplier Thermo Scientific™ Supplier No. U400X
Estimated Shipment: 06-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Maintain the performance of your Thermo Scientific™ Valco™ Injector for HPLC with these syringe ports, ferrules, and nuts.
Maintain the performance of your Thermo Scientific™ Valco™ Injector for high-performance liquid chromatography (HPLC) with these Valco HPLC Injector Accessories, which include syringe ports, ferrules, and nuts.
  • Designed for 10-32 port configurations
  • Burr and contaminant free
TRUSTED_SUSTAINABILITY
Specifications
Type High Pressure Male Hex Nut
Description Male Nut
For Use With (Equipment) 1/16” tubing
Material Stainless Steel (316)
Quantity 10 Pack
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.