missing translation for 'onlineSavingsMsg'
Learn More

Valdecoxib-13C2,15N, TRC

Product Code. 30456154
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456154 10 mg -
30448972 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30456154 Supplier Toronto Research Chemicals Supplier No. TRCV09000210MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1189428-23-6
Molecular Formula 13C2 C14 H14 15N N O3 S
Molecular Weight (g/mol) 317.34
Synonym 4-[3-Phenyl-5-(methyl-13C)isoxazol-4-yl-5-13C-15N]benzenesulfonamide, 4-[5-(Methyl-13C)-3-phenyl-4-isoxazolyl-5-13C-2-15N]benzenesulfonamide, Valdecoxib-13C2,15N, Valdecoxib-13C2,15N (5-(methyl-13C)isoxazol-5-13C-15N)
IUPAC Name 4-(5-(1^{13}C)methyl-3-phenyl-(5^{13}C,2^{15}N)1,2-oxazol-4-yl)benzenesulfonamide
SMILES [13CH3][13c]1o[15n]c(c2ccccc2)c1c3ccc(cc3)S(=O)(=O)N

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C16H14N2O3S/c1-11-15(12-7-9-14(10-8-12)22(17,19)20)16(18-21-11)13-5-3-2-4-6-13/h2-10H,1H3,(H2,17,19,20)/i1+1,11+1,18+1
Formula Weight 317.0763
Chemical Name or Material Valdecoxib-13C2, 15N
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.