missing translation for 'onlineSavingsMsg'
Learn More

Valdecoxib Dimer, TRC

Product Code. 30454272
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30454272 10 mg 10mg
30457119 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30454272 Supplier Toronto Research Chemicals Supplier No. TRCV09002010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1373038-60-8
Molecular Formula C32H25N3O6S2
Molecular Weight (g/mol) 611.69
Synonym N-[4-(5-Methyl-3-phenyl-4-isoxazolyl)benzenesulfonyl]-4-(5-methyl-3-phenyl-4-isoxazolyl)benzenesulfonamide, Valdecoxib Impurity B
SMILES Cc1onc(c2ccccc2)c1c3ccc(cc3)S(=O)(=O)NS(=O)(=O)c4ccc(cc4)c5c(C)onc5c6ccccc6

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C32H25N3O6S2/c1-21-29(31(33-40-21)25-9-5-3-6-10-25)23-13-17-27(18-14-23)42(36,37)35-43(38,39)28-19-15-24(16-20-28)30-22(2)41-34-32(30)26-11-7-4-8-12-26/h3-20,35H,1-2H3
Chemical Name or Material Valdecoxib Dimer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.