Learn More
Valeramide, 97%
Product Code. 11383606
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11383606 25 g 25g
11373606 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11383606 Supplier Thermo Scientific Alfa Aesar Supplier No. L14601.14
 Restricted Item
In Stock
Add to basket
missing translation for 'validMainframeMsg'
CAS: 626-97-1 | C5H11NO | 101.149 g/mol
Valeramide is used in r GC (Gas Chromatography) and LC (Liquid Chromatography) analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Valeramide is used in r GC (Gas Chromatography) and LC (Liquid Chromatography) analysis.
Solubility
Soluble in water (almost transparency).
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 626-97-1
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00041895
InChI Key IPWFJLQDVFKJDU-UHFFFAOYSA-N
Synonym valeramide, n-valeramide, pentanimidic acid, unii-tkl9tn43qk, tkl9tn43qk, ksc353i5p, ipwfjlqdvfkjdu-uhfffaoysa, valeramide, pharmaceutical impurity standard
PubChem CID 12298
ChEBI CHEBI:16459
IUPAC Name pentanamide
SMILES CCCCC(=O)N
Specifications
Melting Point 102°C to 104°C
Quantity 25 g
Beilstein 1740792
Solubility Information Soluble in water (almost transparency).
Formula Weight 101.15
Percent Purity 97%
Chemical Name or Material Valeramide