missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-Valerianol, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | (+)-Valerianol |
| CAS | 20489-45-6 |
| Molecular Formula | C15 H26 O |
| Quantity | 50 mg |
| Synonym | (2R,8R,8aS)-1,2,3,4,6,7,8,8a-Octahydro-α,α,8,8a-tetramethyl-2-naphthalenemethanol (ACI), 2-Naphthalenemethanol','1,2,3,4,6,7,8,8a-octahydro-α,α,8,8a-tetramethyl-','[2R-(2α,8β,8aβ)]- (ZCI), 4βH,5α-Eremophil-1(10)-en-11-ol (8CI), (+)-Valerianol, 2-((2R,8R,8AS)-8,8a-dimethyl-1,2,3,4,6,7,8,8a-octahydronaphthalen-2-yl)propan-2-ol, Kusenol, Kusunol, Valerianol |
| InChI Formula | InChI=1S/C15H26O/c1-11-6-5-7-12-8-9-13(14(2,3)16)10-15(11,12)4/h7,11,13,16H,5-6,8-10H2,1-4H3/t11-,13?,15+/m1/s1 |
| SMILES | C[C@@H]1CCC=C2CCC(C[C@@]12C)C(C)(C)O |
| IUPAC Name | 2-[(8R,8aS)-8,8a-dimethyl-2,3,4,6,7,8-hexahydro-1H-naphthalen-2-yl]propan-2-ol |
| Molecular Weight (g/mol) | 222.366 |
| Formula Weight | 222.198 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction