missing translation for 'onlineSavingsMsg'
Learn More

Valeryl Chloride, TRC

Product Code. 30449268
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30449268 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449268 Supplier Toronto Research Chemicals Supplier No. TRCV09146010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 638-29-9
Molecular Formula C5H9ClO
Quantity 10 g
Synonym Pentanoic Acid Chloride, Valeroyl Chloride, n-Butanecarbonyl Chloride, n-Pentanoyl Chloride
InChI Formula InChI=1S/C5H9ClO/c1-2-3-4-5(6)7/h2-4H2,1H3
SMILES CCCCC(=O)Cl
IUPAC Name pentanoyl chloride
Molecular Weight (g/mol) 120.58
Formula Weight 120.0342
Chemical Name or Material Valeryl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.