missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Validamycin A (>70%) Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Validamycin A Hydrochloride Salt |
| Molecular Formula | C20 H35 N O13 . x[HCl] |
| Quantity | 100 mg |
| Synonym | 1,5,6-Trideoxy-4-O-beta-D-glucopyranosyl-5-(hydroxymethyl)-1-[[(1S,4R,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)-2-cyclohexen-1-yl]amino]-D-chiro-inositol Hydrochloride Salt, Cepex 10SL Hydrochloride Salt, Jinganmycin A Hydrochloride Salt, Jinggang Hydrochloride Salt, Jinggangmycin Hydrochloride Salt, Jinggangmycin A Hydrochloride Salt, Validacin Hydrochloride Salt, Validamycin Hydrochloride Salt, Validamycin A Hydrochloride Salt, [1S-(1alpha,4alpha,5beta,6alpha)]-1,5,6-Trideoxy-4-O-β-D-glucopyranosyl-5-(hydroxymethyl)-1-[[4,5,6-trihydroxy-3-(hydroxymethyl)-2-cyclohexen-1-yl]amino]-D-chiro-inositol Hydrochloride Salt |
| InChI Formula | InChI=1S/C20H35NO13.ClH/c22-3-6-1-8(12(26)15(29)11(6)25)21-9-2-7(4-23)19(17(31)13(9)27)34-20-18(32)16(30)14(28)10(5-24)33-20;/h1,7-32H,2-5H2;1H/t7-,8+,9+,10-,11-,12+,13+,14-,15+,16+,17-,18-,19-,20+;/m1./s1 |
| SMILES | OCC1=C[C@@]([H])([C@@H]([C@H]([C@@H]1O)O)O)N[C@@H]2[C@@H]([C@H]([C@@H]([C@H](C2)CO)O[C@@]3(O[C@@H]([C@H]([C@@H]([C@H]3O)O)O)CO)[H])O)O.[HCl] |
| IUPAC Name | (2R,3R,4S,5S,6R)-2-[(1R,2R,3S,4S,6R)-2,3-dihydroxy-6-(hydroxymethyl)-4-[[(1S,4R,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)cyclohex-2-en-1-yl]amino]cyclohexoxy]-6-(hydroxymethyl)tetrahydropyran-3,4,5-triol;hydrochloride |
| Molecular Weight (g/mol) | 533.95174 |
| Formula Weight | 533.1875 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?