Learn More
Thermo Scientific™ NanoDrop™ Cuvette Qualification Filter Kit
Product Code. 31921092 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
NanoDrop UltraC, NanoDrop UltraC FL, NanoDrop OneC, and NanoDrop 2000C Spectrophotometers
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
31921092 NanoDrop UltraC, NanoDrop UltraC FL, NanoDrop OneC, and NanoDrop 2000C Spectrophotometers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31921092 Supplier Thermo Scientific™ Supplier No. 840257300
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Cuvette Qualification Filter Kit for the cuvette based Thermo Scientfic™ NanoDrop™ UltraC and NanoDrop™ UltraC FL, NanoDrop™ OneC and NanoDrop™ 2000c Spectrophotometers is an essential tool for ensuring the optimal performance of your spectrophotometers. This kit provides you with the necessary filters for accurate cuvette qualification testing.
Includes: 1 x 10%T Filter (Calibrated), 1 x 50%T (Calibrated), and a certificate with traceability and NVLAP 17025, Lab Code 200955, accreditation.
Specifications
Description Qualification Filter Kit for NanoDrop OneC and NanoDrop 2000C Spectrophotometers
For Use With (Equipment) NanoDrop UltraC, NanoDrop UltraC FL, NanoDrop OneC, and NanoDrop 2000C Spectrophotometers
Includes 1 x Filter holder blank, 1 x 10%T Filter, Calibrated, 1 x 50%T Filter, Calibrated, Certificate with traceability and NVLAP 17025, Lab Code 200955, accreditation