missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Valsartan, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Valsartan |
| CAS | 137862-53-4 |
| Analyte or Component Names | Valsartan |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H29 N5 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Valsartan, (2S)-3-Methyl-2-[pentanoyl[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]amino]butanoic acid, L-Valine, N-(1-oxopentyl)-N-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]- (9CI), N-(1-Oxopentyl)-N-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-L-valine, CGP 48933, Cardopan, Diovan, Nisis, Tareg, Valsarect, Valsartanin |
| InChI Formula | InChI=1S/C24H29N5O3/c1-4-5-10-21(30)29(22(16(2)3)24(31)32)15-17-11-13-18(14-12-17)19-8-6-7-9-20(19)23-25-27-28-26-23/h6-9,11-14,16,22H,4-5,10,15H2,1-3H3,(H,31,32)(H,25,26,27,28)/t22-/m0/s1 |
| Solution Type | Neat |
| SMILES | CCCCC(=O)N(Cc1ccc(cc1)c2ccccc2c3nnn[nH]3)[C@@H](C(C)C)C(=O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?