missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(III) 2,4-pentanedionate, typically 97%

Product Code. 11368217
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11368217

Brand: Thermo Scientific Alfa Aesar 022940.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13476-99-8 | C15H21O6V | 348.27 g/mol

Vanadium(III) 2,4-pentanedionate is used as homogeneous catalyst, primary and secondary intermediates, Laboratory chemicals, and manufacture of substances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13476-99-8
Molecular Formula C15H21O6V
Molecular Weight (g/mol) 348.27
MDL Number MFCD00000033
InChI Key ZVFSRLXFKIWGRT-UHFFFAOYSA-N
Synonym Vanadium(III) acetylacetonate
PubChem CID 122198782
IUPAC Name (Z)-4-hydroxypent-3-en-2-one;vanadium
SMILES [V+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Quantity 100 g
UN Number UN3285
Sensitivity Air Sensitive
Solubility Information Soluble in water 2g/L,toluene,ethanol,acetone.
Formula Weight 348.27
Percent Purity 97% (typically)
Physical Form Powder
Chemical Name or Material Vanadium(III) 2,4-pentanedionate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.