missing translation for 'onlineSavingsMsg'
Learn More

Vanilloid R1/TRPV1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications

213.00€

Specifications

Antigen Vanilloid R1/TRPV1
Concentration 1 mg/mL
Dilution Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen 1 μg/mL
Applications Western Blot
Classification Polyclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18594584
View Documents
Novus Biologicals
NB120-3487-0.025mg
0.025 mg
213.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Vanilloid R1/TRPV1 Polyclonal specifically detects Vanilloid R1/TRPV1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications

Specifications

Vanilloid R1/TRPV1
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen 1 μg/mL
Polyclonal
Rabbit
Neuroscience, Sensory Systems
PBS, 1 mg/ml BSA with 0.05% Sodium Azide
Capsaicin receptor, Osm-9-like TRP channel 1, OTRPC1, transient receptor potential cation channel subfamily V member 1, transient receptor potential cation channel, subfamily V, member 1, transient receptor potential vanilloid 1a, transient receptor potential vanilloid 1b, TrpV1, Vanilloid receptor 1, vanilloid receptor subtype 1, VR1, VR1DKFZp434K0220
TRPV1
IgG
Affinity Purified
Not yet tested in other species.
1 mg/mL
Western Blot
Unconjugated
RUO
Human
Q52PU4
7442
Synthetic peptide corresponding to residues T(7) D L G A A A D P L Q K D T C(21) of human VR1.
Primary
Store at -20°C. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.