Learn More
VAPA Mouse, anti-Human, Unconjugated, Clone: 1F10F9, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67845-1-IG-150UL
 View more versions of this product
Product Code. 17281506
  • 410.00€ / 150µL
Description
VAPA (Vesicle-associated membrane protein-associated protein A), also known as VAP33, is a 33-kDa, ubiquitously expressed integral membrane protein of the VAMP-associated protein (VAP) family. It is present in the plasma membrane and in intracellular vesicles, playing a role in vesicle trafficking.
Specifications
Specifications
VAPA
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat
Western Blot, Immunohistochemistry, ELISA
1F10F9
Unconjugated
150 μL
9218