Learn More
VAPA Mouse, anti-Human, Unconjugated, Clone: 1F10F9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67845-1-IG-20UL
Description
VAPA (Vesicle-associated membrane protein-associated protein A), also known as VAP33, is a 33-kDa, ubiquitously expressed integral membrane protein of the VAMP-associated protein (VAP) family. It is present in the plasma membrane and in intracellular vesicles, playing a role in vesicle trafficking.
Specifications
| VAPA | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat |
| Western Blot, Immunohistochemistry, ELISA | |
| 1F10F9 | |
| Unconjugated | |
| 20 μL | |
| 9218 | |
| Liquid |