missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VAPA Recombinant Rabbit Monoclonal Antibody (24GB2580)

Rabbit Recombinant Monoclonal Antibody
198.00€ - 564.00€
Specifications
| Antigen | VAPA |
|---|---|
| Clone | 24GB2580 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275278
![]() |
Invitrogen™
MA557836 |
100 μL |
564.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30276243
|
Invitrogen™
MA557837 |
20 μL |
198.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VAPA Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The protein encoded by this gene is a type IV membrane protein. It is present in the plasma membrane and intracellular vesicles. It may also be associated with the cytoskeleton. This protein may function in vesicle trafficking, membrane fusion, protein complex assembly and cell motility. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.Specifications
| VAPA | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| VAPA | |
| 33 kDa VAMP-associated protein; 33kDa; hVAP-33; synaptobrevin-associated protein of 33kDa; vamp; VAMP (vesicle-associated membrane protein)-associated protein A; VAMP (vesicle-associated membrane protein)-associated protein A, 33kDa; VAMP associated protein A; VAMP-A; VAMP-associated protein A; VAP33; VAP-33; Vapa; VAP-A; vesicle-associated membrane protein, associated protein a; vesicle-associated membrane protein-associated protein A | |
| VAPA | |
| Primary | |
| Affinity chromatography |
| 24GB2580 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9P0L0, Q9WV55, Q9Z270 | |
| 30960, 58857, 9218 | |
| A synthesized peptide derived from human VAPA (aa 10-58). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title